Would you like to log in again?
Pocket Steel Rules, Mitutoyo
Supplier: Mitutoyo
504-950-300
300049-054EA 18.71 USD InStock
300049-054
Pocket Steel Rules, Mitutoyo
Rulers
Pocket steel rules are easy to read.
  • Pocket clip
Resolution: ¹/₆₄, 1 mm
Accuracy: +0.004"/−0.0035"
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty