Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-B. burgdorferi antigen Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: SeraCare
5330-0064
95058-200EA 497.44 USD InStock
95058-200
Anti-B. burgdorferi antigen Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Primary Goat Anti-B. burgdorferi Reacts with Borrelia burgdorferi

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Borrelia burgdorferi

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty