Would you like to log in again?
Cart Sprayers, Chapin™
Supplier: Chapin™
139-6300
300091-574EA 771.3 USD InStock
300091-574
Cart Sprayers, Chapin™
Spray Guns
Stainless steel cart sprayer with D-handle.
  • Adjustable nozzle
  • 18" extension
  • 10 ft. hose length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty