Would you like to log in again?
SpectraMax® 190 Absorbance Plate Reader, Molecular Devices
Supplier: Molecular Devices
SpectraMax®
190-G 190-SC 0310-3268
10067-758EA 60581.85 USD InStock
10067-758 89212-394 76358-622
SpectraMax® 190 Absorbance Plate Reader, Molecular Devices
Microplate Readers
Enables data acquisition in endpoint, kinetic, and spectral scan modes at wavelengths across the UV-visible wavelength range, with the only temperature-independent method of pathlength correction.
  • Temperature control
  • Shaking capabilities
  • Multichannel optical design
  • Eliminates error
Temperature control and shaking come standard.
The patented multichannel optical design of the SpectraMax® 190 Absorbance Microplate Reader mimics a dual beam spectrophotometer, measures each sample in the plate directly, and eliminates measurement error due to variations in light output between optical fibers on other instruments for more reliable data reporting. The eight-channel system delivers both high precision and speed when reading 96-well microplates for endpoint measurements, kinetic assays, and spectral scans. Narrow, 2nm bandwidths deliver increased measurement accuracy and linearity over the widest range of assays, including ELISAs in the visible range or DNA quantitation in the UV range.
The SpectraMax® 190 can read as low as 2μL with the μMax Low Volume Plate.
Ordering information: Includes current version of SoftMax Pro Software. Additional models are available. Additional accessories are available including the µMax Low Volume Plate, standard validation tools, and 21 CFR Part 11 Compliance tools. Standard 12-month warranty included. Purchase of Additional 1 Year Service Plan includes: MRC Essential Service Plan, including parts, labor, and shipping/handling costs for emergency repairs for one year at the Molecular Devices Repair Center. Priority repair scheduling, along with unlimited technical support and loaner instrument (if available), are provided with this service option.
Order Now
ORDER
SPECIFICATIONS
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty