Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Vacuum Trap Assembly, Ace-Thred Top and Inlet/Outlet, Easy-Action Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
8748-47
89063-890EA 2079.17 USD InStock
89063-890
Vacuum Trap Assembly, Ace-Thred Top and Inlet/Outlet, Easy-Action Stopcock, Ace Glass Incorporated
Vacuum Traps
This twin chambered dry ice vacuum trap is designed to be used with rotary evaporators or any other large solvent evaporation applications
The inlet/outlet ports have been modified with #15 Ace-Threds for use with PTFE "Ace-Safe" tubing connectors to make an easy, leak-tight, safe connect/disconnect to tubing. These ports are located on the sides of chambers; one is for connection to the evaporator, the other to the vacuum source. Both are for use with 1/2-inch I.D. tubing. The stopcock at bottom is 0-5mm Easy-Action threaded style with double PTFE ring seals on plug, for draining both chambers simultaneously. Normally, the condensate will collect in the chamber connected nearest to the evaporator until it is filled to crossover between chambers. Then the second chamber will fill. The capacity of the vacuum traps is 450mL each. The capacity of the chambers with the traps inserted is approximately 600mL each. Overall height of the unit is approximately 20 inches.
The unique, squatty design is 18 inches high, and the use of twin chambers offers increased condensing capacity that would be attained only with a much taller, single chambered condenser. The vacuum traps are secured into the main body chambers via #50 Ace-Threds using bushings and o-rings. This type of connection allows for easy removal for cleaning. Fabricated from borosilicate glass.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty