Would you like to log in again?
Wellwash® Microplate Washer, Thermo Scientific
Supplier: Thermo Scientific
5165000
97048-766EA 11962.28 USD InStock
97048-766
Wellwash® Microplate Washer, Thermo Scientific
The Wellwash® Microplate Washer is an easy-to-use strip washer designed for use with 96-well plates in routine ELISA applications
A context specific help feature ensures that protocols are developed quickly and easily, with minimal user training required. The Wellwash® Microplate Washer is designed to provide accurate and precise dispensing and aspiration of washing solutions via specially designed wash heads. The washing parameters are easily adjustable to guarantee the best possible performance. This strip washer incorporates a sweep mode to ensure extremely low residual volumes. Additionally, each unit features a USB port for transferring protocols between several washers, and for printing reports and logfiles.
The unit features a large color display with an intuitive graphical user interface, which makes each assay extremely easy to set up. The internal software is available in nine different languages (English, French, German, Italian, Spanish, Portuguese, Russian, Japanese, and Chinese).
Ordering information: Wellwash® Microplate Washers are supplied with one 2L non-pressurized wash bottle and one 2L waste bottle. Units are not intended for cell washing.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty