Would you like to log in again?
GC-8IF Gas Chromatograph, Shimadzu
Supplier: Shimadzu
221-23354-92
89217-148EA 15572.07 USD InStock
89217-148
GC-8IF Gas Chromatograph, Shimadzu
Chromatography Systems Gas Chromatographs
The isothermal GC-8A with dual, differential FID (Flame Ionization detector) incorporates an uncomplicated, rugged design making it ideal for dedicated applications and simple to operate
Two on-column injection ports, differential flow control, compact design and rugged construction make this GC ideal for high volume dedicated use.
Ordering information: Installation included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty