Would you like to log in again?
Dust Cap Drum Cover, New Pig
Supplier: New Pig
DRM149
56620-778CS 448.88 USD InStock
56620-778
Dust Cap Drum Cover, New Pig
Covers
Block dirt and moisture out of containers to keep contents pure
  • Durable, low-density polyethylene
  • Meets FDA requirements
  • Recommended for indoor use
Clear 4 mil polyethylene protects contents but makes them easy to inspect.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty