Would you like to log in again?
Galileo Kit
Supplier: Avantor
WARD470152-832
470152-832EA 69.99 USD InStock
470152-832
Galileo Kit
Educational Classroom Kits and Activities
Re-create His Famous Experiment
  • Includes an inclined plane, steel ball, funnel assembly, and instructions
  • A graduated cylinder, support stand, and clamp, required, are available separately
Galileo discovered, using an inclined plane and a ball, that objects fall at a constant rate of acceleration; if the angle of the inclined plane is changed, it does not affect that rate. Now you and your students can observe the same effect in your own classroom.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty