Would you like to log in again?
Firefly Gene Transformation Kit
Supplier: Avantor
470007-118KT 149.09 USD InStock
470007-118
Firefly Gene Transformation Kit
Educational Classroom Kits and Activities
This kit is a dramatic test of a novel reporter gene.
  • Bacteria glows without a UV light
  • Pre-poured petri plates included
  • Links to real-world concepts
  • Estimated required lab time: 45 minutes over two or three days
Bacteria that glow in the dark? It's possible with our lab activity. Students perform true gene transfer between two completely different types of organisms: a firefly and a bacterium. We provide you with the bacteria and plasmid DNA with the luminescence gene from a firefly.
Students transfer the firefly gene to the bacteria and employ a chemical substrate to make the bacteria glow. Dimmed lights work just as well as an ultraviolet light.
Ordering information: This kit includes capillary micropipettes, nitrocellulose membranes, sterile inoculating loops, microcentrifuge tubes, pipette bulbs, transfer pipets, Luria agar, sodium citrate, Luria broth, and calcium chloride. Live and perishable materials include E.coli, Plasmid pBEST-Luc, Luciferin, and LB Amp plates. The kits also includes a coupon for redemption of perishable materials. Redeem by mail, phone, fax, or email. A hot-water bath or hot plate, beakers, colored candies, and a black bag are needed but not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty