Would you like to log in again?
Kestrel USB Interface Package
Supplier: Kestrel Meters
0804USB
470021-642EA 164.05 USD InStock
470021-642
Kestrel USB Interface Package
Weather Stations
Analyze your adventures after the fact with this convenient system.
All data gathered during your field studies can be uploaded for long term storage, in-depth analysis, and detailed charting. Simply connect this interface cable to a computer’s USB port and the Kestrel communicator automatically recognizes the model of meter being used, then uploads the data. Package includes interface cradle unit, USB cable, and CD with upload software.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty