Would you like to log in again?
Preserved Lamprey
130.1022.10 130.1022.1 130.1014.10 130.1006.1 130.1006.10
470001-746EA 19.99 USD InStock
470001-746 470001-454 470001-458 470001-744 470001-456
Preserved Lamprey
Specimens Preserved Specimens Preserved Animal Materials
Formalin preserved lamprey. Available in vacuum packs.
  • Fish anatmoy
  • Agnathan anatomy
  • Evolutionary biology
Petromyzon marinus. Representative of the Agnathans or jawless cartilaginous fish. Lampreys use their suction cup-like mouth to attach to their host and then rasp away the host fish’s skin to feed on its blood and bodily fluids. A major pest in the United States’ Great Lakes, lamprey have greatly depleted the populations of other fish. Chemical control of lamprey larvae has reduced their population to a tolerable level.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty