Would you like to log in again?
Seasonal Display Globe
Supplier: THE SCIENCE SOURCE SE
U010
470004-370EA 2371.4 USD InStock
470004-370
Seasonal Display Globe
Globes
A Captivating Way to Show the Relationship Between the Sun and the Seasons
Observe how the amount and angle of sunlight cast on our planet changes during the year, causing day and night, and the different seasons. The handcrafted globe is contained in a plexiglass cover that re-creates the blackness of space. Lit from within, the globe is shielded so that only half the planet is visible as it turns, showing how the sun illuminates half the Earth at a time. The shield gradually rotates over a six minute period to simulate the changing pattern of sunlight falling over each hemisphere during the year. A bright spot represents the position on the Earth where the sun is directly overhead. The multi-setting control allows you to observe the globe with lit rotation, unlit rotation, or no rotation. Some assembly required. 110 VAC; Globe size: 16" dia.; plexiglass cover size: 18"L x 18"W x 23"H.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty