Would you like to log in again?
Vernier® Microphone
Supplier: VERNIER SOFTWARE & TECH SE
MCA-BTA
470002-962EA 54.99 USD InStock
470002-962
Vernier® Microphone
Sound Meters
Vernier Microphone helps students learn about waveforms of sound.
  • For Displaying and Studying Waveforms
  • Typical Maximum Frequency: 10,000 Hz
  • Power: 1.45 mA At 5 VDV
  • Ideal for Speed of Sound Experiments
  • For Educational Use Only
The Microphone can be used to display and study the waveforms of sounds from voices and musical instruments.
It is also great for speed of sound experiments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty