Would you like to log in again?
SMITH & WESSON® 44 MAGNUM™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Smith & Wesson®
23454 23453 23452 23457
89130-966EA 7.68 USD InStock
89130-966 89130-962 89130-964 89130-960
SMITH & WESSON® 44 MAGNUM™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Safety Glasses
Soft-touch temples naturally contour the head for a secure and comfortable fit.
  • Temple vents maximize airflow and minimize fogging in humid conditions
  • Cut-to-shape distinctive and stylish one-piece lens has a 7-base curve
Floating, flexible nose piece easily conforms to various nose shapes and sizes for additional comfort and flexibility. Scratch-resistant polycarbonate lens provides 99.9% U.V. protection.
Certifications: Meets ANSI Z87.1+ standards and is CSA Z94.3 compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty