Would you like to log in again?
PCR Grade dNTPs, Biosearch Technologies
Supplier: Biosearch Technologies
30030-1 30029-1
89137-892EA 251.74 USD InStock
89137-892 89137-886
PCR Grade dNTPs, Biosearch Technologies
Lucigen's PCR Grade dNTPs are the highest quality (> 99% purity) deoxynucleotides, 2'-deoxynucleoside 5'-triphosphates
These nucleotides are suitable for: 1) Sequencing, 2) Nick translation, 3) cDNA synthesis, 4) TdT-tailing reactions, 5) Fill-in reactions. 2.5 mM dNTP Mix, PCR Grade:
This ready-to-use dNTP Mix is a mixture of all four nucleotides (dATP, dCTP, dGTP, dTTP), each dissolved in highly purified water at a concentration of 2.5 mM, at pH 8.3. Two milliliters of the Mix is enough for 500 standard 50 µl PCR reactions.
10 mM dNTP Set, PCR Grade:
This high-purity dNTP set allows you to formulate your own PCR mix. Each of all four nucleotides (dATP, dCTP, dGTP, dTTP), is provided in a separate vial (750 µl each), dissolved at a concentration of 10 mM in highly purified water, pH 8.3.
They are formulated for your convenience and come at an excellent value.
Ordering information: The 2.5 mM dNTP Mix is a mixture of all four nucleotides, each dissolved in highly purified water at a concentration of 2.5 mM, at pH 8.3. The 10 mM dNTP Set provides each nucleotide in a separate vial (750 µl each), dissolved at a concentration of 10 mM in highly purified water, pH 8.3. Please Note: Some applications in which Lucigen’s dNTPs can be used may be covered by patents issued and applicable in the United States and certain other countries. Because purchase of this product does not include a license to perform any patented application, users of this product may be required to obtain a patent license depending upon the particular application in which the product is used. The PCR process is the subject of European Patent Nos. 201,184 and 200,262 owned by Hoffman-LaRoche. Those patents expired on March 28, 2006. The corresponding PCR process patents in the United States expired on March 29, 2005. It is the sole responsibility of the buyer to ensure that use of the product does not infringe the patent rights of third parties.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty