Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylindoline ≥98.0%

Supplier: TCI America
Warning

M0871-25ML
TCM0871-025MLEA 55.8 USD InStock
TCM0871-025ML
2-Methylindoline ≥98.0%
2-Methylindoline
CAS Number: 6872-06-6
MDL Number: MFCD00005706
Molecular Formula: C9H11N
Molecular Weight: 133.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 93
Specific Gravity (20/20): 1.02


Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 230 °C
Density: 1.02 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00005706
CAS Number: 6872-06-6
EINECS: 229-971-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/75/28974375.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty