Synonyms:
1-Isobutyl-1H-imidazo[4,5-c]quinolin-4-amine
, 4-Amino-1-isobutyl-1H-imidazo[4,5-c]quinoline
I0747-1G
I0747-100MG
TCI0747-100MGEA
67.99
USD
InStock
TCI0747-100MG
TCI0747-1G
Imiquimod ≥98.0% (by HPLC, titration analysis)
Imiquimod
CAS Number: 99011-02-6
MDL Number: MFCD00866946
Molecular Formula: C14H16N4
Molecular Weight: 240.31
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 294
Lambda max.: 245 nm (H2O/MeOH)
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Antitumor Ingredients, Antiviral Ingredients
Other Product Names / Synonyms: 1-Isobutyl-1H-imidazo[4,5-c]quinolin-4-amine, 4-Amino-1-isobutyl-1H-imidazo[4,5-c]quinoline