Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Sitosterol 40,0% (contains Campesterol)

Supplier: TCI America
Synonyms: α-Dihydrofucosterolβ-Sitosterol24α-Ethylcholesterol22,23-Dihydrostigmasterol5-Stigmasten-3β-ol

S0040-25G
TCS0040-025GEA 75.09 USD InStock
TCS0040-025G
β-Sitosterol 40,0% (contains Campesterol)
beta-Sitosterol
CAS Number: 83-46-5
MDL Number: MFCD00003631
Molecular Formula: C29H50O
Molecular Weight: 414.72
Purity/Analysis Method: 40.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 140
Specific rotation [a]20/D: -35 deg (C=2, Dioxane)


Formula: C₂₉H₅₀O
MW: 414.72 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00003631
CAS Number: 83-46-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/39/12/28993912.jpg
Order Now
Specification Test Results
Purity/Analysis Method 40.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty