Would you like to log in again?
dNTP Mixture 100 mM Solution, OmniPur®, MilliporeSigma
Supplier: MilliporeSigma
Calbiochem®
3640-1PC
EM-3640EA 206.5 USD InStock
EM-3640
dNTP Mixture 100 mM Solution, OmniPur®, MilliporeSigma
Nucleic Acid Reagents Nucleotides
Ready to use
The dNTP Mix contains ultrapure dATP, dCTP, dGTP, and dTTP (monosodium salts) in sterile deionized water at pH 7.0.
Ideal for any application that requires pure deoxynucleotides, such as PCR, cDNA synthesis, and fill-in reactions.
Exp 12 months from mfg date
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty