Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TF Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-034
RL600-401-034EA 1290 USD InStock
RL600-401-034
Anti-TF Rabbit Polyclonal Antibody
Antibodies
This product has been assayed against 1.0 µg of Human Transferrin in a standard sandwich ELISA using Peroxidase conjugated Affinity Purified Donkey anti-Rabbit IgG (H&L) MX7 and TMB as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:500 - 1:2500;

Type: Primary
Antigen: TF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty