Would you like to log in again?
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 200 mesh woven from 0.05mm (0.002in) Type 304
Supplier: Thermo Scientific Chemicals
013476-CH 13476-RU 13476-RR 013476-RR 13476-CH 013476-RU
AA13476-RREA 45.19 USD InStock
CAAA13476-RR CAAA13476-RU AA13476-RR CAAA13476-CH AA13476-RU AA13476-CH
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 200 mesh woven from 0.05mm (0.002in) Type 304
Stainless steel
Open area: 36%; Width of opening: 0.003in
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty