Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Motic DSK-500 Binocular Stereo Microscope, Motic Instruments
Supplier: Motic
1100200900011 1100200900011
19000-212EA 5920.9 USD InStock
19000-212 CA19000-212
Motic DSK-500 Binocular Stereo Microscope, Motic Instruments
Microscopes
Offering the benefits of a Common Main Objective [CMO] infinity optical system at a Greenough price, the K500 is the ideal choice for users wanting the flatness and expandability of a CMO but only possessing a Greenough budget.
The K500 utilises a 5-step magnification changer with industry standard stops at 0.64X, 1.0X, 1.6X, 2.5X, and 4.0X for convenience in converting from existing systems for inspection and observation applications. The system can be converted from a simple optical inspection/observation system into a documentation and analysis station with the attachment of the 1500.3 Video/Photomicrography optical bridge and the user's choice of documentation [photomicrography or digital].
Employing a comfortable viewing observation angle of 45,° the K500’s ergonomic layout is further complemented by the side location of the step magnification changer for rapid magnification changes. Moreover, the guaranteed parfocality of throughout the system combined with the size of the K500 makes the decision to switch over to a CMO system hassle-free.
The K500 is available also in a dual discussion system, DSK-500, for applications were you want to promote discussion or need a second option quickly.
The K500 utilises a 5-step magnification changer with industry standard stops at 0.64X, 1.0X, 1.6X, 2.5X, and 4.0X for convenience in converting from existing systems for inspection and observation applications
Certifications: UL Listed.
Ordering information: Microscope is supplied with all accessories listed above. Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a 1 year electrical and 5 year mechanical warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty