Would you like to log in again?
Helmholtz Coils Apparatus
Supplier: VWR International
PH0845HEL
470121-440EA 162.2 USD InStock
470121-440
Helmholtz Coils Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate superposition of magnetic fields with this easy-to-use apparatus.
  • Each coil contains 400 turns of copper wire, wound on former of approx. 150 mm diameter
  • 4 mm 'banana plug' terminals
  • Maximum current in the coil is 1 A to avoid overheating
  • Coil separation can range from approx. 1.75 to 12"
  • Direction of current is marked on each coil
A pair of precision coils and 400 turns of wire are wound on a heat-resistant former of approximately 150 mm diameter. The coil is comprised of a support rod, which fits on the holders of the track system supplied, enabling them to be aligned on a common axis for measurements and study of the magnetic fields.
Normally used in pairs, each coil has 4 mm terminals and their distance apart is adjusted over the track supplied. The maximum current in the coil is 1 A to avoid overheating. Direction of current is marked on each coil. The complete apparatus measures 6.5" tall , 11" long, and 6.5" wide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty