Would you like to log in again?
C Columns, Empty Chromatography Columns
Supplier: Cytiva
19-5203-01 19-5104-01 19-5201-01 19-5101-01 19-5203-01 19-5101-01 19-5104-01 19-5201-01 19-5001-01 19-5102-01 19-5002-01 19-5103-01 19-5001-01 19-5003-01 19-5002-01 19-5102-01 19-5202-01 19-5003-01 19-5103-01 19-5202-01
10000-300EA 437.69 USD InStock
10000-300 10000-288 10000-044 10000-298 10000-296 10000-042 10000-294 10000-040 10000-292 CA10768-738 10000-290 CA10768-748 CA10768-750 CA10768-752 CA10768-400 CA10768-740 CA10768-754 CA10768-742 CA10768-756 CA10768-744
C Columns, Empty Chromatography Columns
Chromatography Columns
C columns are economical empty columns suitable for low pressure liquid chromatography and gravity feed at the lower volumes used at research scale.
  • Economical – for low pressure liquid chromatography at research scale
  • Broad compatibility – functions with a stand-alone pump, chromatography system, or gravity feed
  • Precise temperature control – with the optional cooling jacket
  • Efficient packing – packing reservoirs are available as accessories specific to each column diameter
Standard C-series columns have a fixed volume, but they can be upgraded through the addition of flow adapters. These work to keep bed height constant and protected from insoluble particles during chromatography. Adjustable flow adapters are available for 10 mm (AC 10), 16 mm (AC 16), and 26 mm (AC 26) diameter C columns.
Three RC packing reservoirs help optimize packing conditions, especially when packing high beds. They are available for each diameter in the C column series, and are quick and easy to assemble. After the resin is in the column, the RC reservoir can be connected to a pump or Mariotte flask to give optimal flow rates for packing.
Resolution and recovery can often be increased by fitting C columns with a JC thermostatic jacket during temperature-dependent low-pressure chromatography. JC jackets increase control of separation temperature and mechanical resistance of these columns, enabling operating pressures of up to 1 bar.
Delivery information: Each C column comes equipped with polyethylene tubing (1.0 mm inner diameter, 2 m length), bed support screens, snap-on net rings, and instructions for research-scale column chromatography use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty