Would you like to log in again?
HiScale Columns, Cytiva
Supplier: Cytiva
HiScale™
28-9645-14 28-9644-24 28-9644-44 28-9644-45 28-9644-44 28-9645-13 28-9644-45 28-9645-13 28-9644-24 28-9644-41 29360550 28-9644-41 28-9645-14
10000-048EA 1297.17 USD InStock
CA10768-770 10000-048 10000-058 10000-056 10000-054 10000-052 10000-060 CA10768-768 76358-650 CA10768-772 CA10768-760 CA10768-764 CA10768-766
HiScale Columns, Cytiva
Chromatography Columns
Empty columns with high pressure stability (able to withstand up to 20 bar of pressure) and high flow rates are compatible for packing with modern BioProcess™ chromatography media, such as MabSelect™ and Capto™ for process development and preparative chromatography. The columns are designed to be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of biomolecules, with the following few exceptions: chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol. Solutions containing more than 2 M NaOH, 1 M HCl, other strong mineral acids or 75% acetic acid should not be used. The column hardware consists of borosilicate glass, PC, PA (reinforced glass fibre), EPDM, POM, PEEK, ETFE, Fluoro-rubber, PP and PA (nylon).
  • Provides axial compression of the gel bed and a non rotating plunger mechanism that expands packing capabilities
  • Dual adapters offer high flexibility and dynamic packing heights
  • Capability to rinse entire adapter shaft after packing minimises the risk of bacterial growth after prolonged use
The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments as well as disassembly and cleaning. The measurement scale on the column makes it easier to predict and reach a predefined packing height, thereby increasing process control as well as reproducibility. Although optimized for high-pressure media, HiScale columns are fully compatible with all applications (and packing methods) currently applicable to XK columns.
The user friendly design allows for an optimized process, through simplified and reproducible operation.
Delivery information: HiScale columns are delivered with a column tube, outer protection tube, two adapters, PEEK tubing, and unions at both ends for easy connection to valves, pumps, and monitors.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty