Would you like to log in again?
APS™ Adhesion Microscope Slides, Matsunami Glass
Supplier: Matsunami Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SUAPS14 SUAPS12 SUAPS1190 SUAPS15 SUAPS11 SUAPS13
10000-324CS 538.41 USD InStock
10000-324 10000-148 10000-322 10000-146 10000-320 10000-144
APS™ Adhesion Microscope Slides, Matsunami Glass
Slides Adhesive Slides
The Matsunami APS™ slide is treated to provide a strong permanent charge to the slide surface
Matsunami microscope slides are manufactured in Japan using optically clear, exceptionally flat, low fluorescence glass. Slides are available with 90° or clipped corners.
Excellent for routine histology, IHC, and cytology preparations. Optimized for use with thermal printers.
Ordering information: Slides are packaged 100 per box, 10 boxes per case (1000 slides).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty