Would you like to log in again?
DuraPro® Steel Toe Boots with Steel Shank, Onguard
Supplier: Onguard
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DuraPro®
89086-15 89086-06 89086-13 89086-10 89086-14 89086-05 89086-07 89086-09 89086-08 89086-16 89086-11 89086-12 89086-03 89086-04
10000-418CS 404.07 USD InStock
10000-418 10000-428 10000-406 10000-416 10000-426 10000-414 10000-424 10000-412 10000-422 10000-410 10000-432 10000-420 10000-430 10000-408
DuraPro® Steel Toe Boots with Steel Shank, Onguard
Shoes and Boots
Protection and comfort is revolutionized with the DuraPro® line of protective footwear.
  • Protective toe cap
  • Comfortable, waterproof protection
  • Anti-skid outsole
  • Fuel oil resistant
  • Chemical resistant
  • Energy absorbing
A 360° pull tab allows for easy donning and doffing, and a sloped comfort top prevents chafing when walking, bending and kneeling. These boots feature a protective toe cap that offers maximum protection against falling objects. The built-in protective toe cap serves to protect from impacts of an energy level of 125 J and a compression at a load of 11121 N (2500 lbf). The boots offer a perfect fit, optimizing comfort for everyday use. They are 100% waterproof to keep feet dry under wet and dirty conditions. The outsole is resistant against oil, which assures that potentially adverse effects of oily substances on the protective properties of the boots are minimized. They also offer protection against a wide variety of chemicals, and feature energy absorption in the heel region which limits impact to the foot while walking. Shock absorption capabilities within the footwear minimizes the risk of injuries.
Certifications: ASTM certified. Steel toe complies with ASTM F2413-11 with steel shank.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty