Would you like to log in again?
DuraPro® XCP Steel Toe Boots with Steel Shank, Onguard
Supplier: Onguard
DuraPro® XCP
84086-09 84086-04 84086-05 84086-07 84086-14 84086-03 84086-06 84086-10 84086-16 84086-08 84086-15 84086-13 84086-11 84086-12
10000-468CS 608.19 USD InStock
10000-468 10000-478 10000-466 10000-488 10000-476 10000-464 10000-486 10000-474 10000-484 10000-462 10000-472 10000-482 10000-470 10000-480
DuraPro® XCP Steel Toe Boots with Steel Shank, Onguard
Shoes and Boots
High performance compound provides ideal chemical resistance in the upper and superior abrasion and slip resistance in the outsole.
  • Protective toe cap
  • Comfortable, waterproof protection
  • 360° pull tab for easy donning
  • Fuel oil resistant
  • Chemical resistant
  • Energy absorbing
A 360° pull tab allows for easy donning and doffing, and a sloped comfort top prevents chafing when walking, bending and kneeling. These boots feature a protective toe cap that offers maximum protection against falling objects. The built-in protective toe cap serves to protect from impacts of an energy level of 125 J and a compression at a load of 11121 N (2500 lbf). The boots offer a perfect fit, optimizing comfort for everyday use. They are 100% waterproof to keep feet dry under wet and dirty conditions. The outsole is resistant against oil, which assures that potentially adverse effects of oily substances on the protective properties of the boots are minimized. They also offer protection against a wide variety of chemicals, and feature energy absorption in the heel region which limits impact to the foot while walking. Shock absorption capabilities within the footwear minimizes the risk of injuries.
Certifications: ASTM certified. Steel toe complies with ASTM F2413-11 with steel shank.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty