Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FLAG Tag Rabbit Polyclonal Antibody (CF594)

Supplier: Biotium

20434
10001-602EA 317.88 USD InStock
10001-602
Anti-FLAG Tag Rabbit Polyclonal Antibody (CF594)
Antibodies
Polyclonal anti-FLAG (DYKDDDDK) reacts with FLAG tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-FLAG IgG (H+L) with CF™594 dye. CF™594 (Ex/Em 593/614 nm) is a deep-red dye spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugates due to its high quantum yield and exceptional water solubility. CF™594 also has excellent photostability ideal for demanding applications, such as confocal microscopy and single molecule imaging.

Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.

Type: Tag
Antigen: FLAG Tag
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Rabbit
Isotype:
Reactivity: FLAG tag

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty