Would you like to log in again?
dNTP Set, Biotium
Supplier: Biotium
40052
10001-638EA 306.96 USD InStock
10001-638
dNTP Set, Biotium
Nucleic Acid Reagents Nucleotides
Highly purified dNTPs (>99% by HPLC) in purified water. Each deoxynucleotide solution (dATP, dCTP, dGTP, dTTP) is provided in a separate vial at 100 mm.
The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty