Would you like to log in again?
dCTP CF® Dye Conjugates, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CF™
40028-T 40027-T 40056 40066 40057 40068-T 40056-T 40027 40058 40066-T 40055 40068 40058-T 40067-T 40028 40067 40055-T 40057-T
76221-638EA 115.22 USD InStock
76221-638 76414-948 10001-646 10098-602 10018-192 10098-600 76221-650 76221-640 10018-194 10001-644 10018-190 10018-188 76221-646 76221-648 76221-652 76221-642 76221-654 76221-644
dCTP CF® Dye Conjugates, Biotium
Protein Linking and Labeling Systems
CF® dye-dCTP can be used to synthesize probes for in situ hybridization, microarray, or to synthesize labeled DNA probes for in situ hybridization and nucleic acid blotting applications.
  • Synthesize exceptionally bright and photostable fluorescent DNA probes
  • Detect by fluorescence microscopy, flow cytometry, or in gel
  • Compatible with fixed cells or tissue sections
  • Choice of nine CF® dye colors from green to far-red
  • Biotiums CF® dyes have superior brightness, photostability, and are water-soluble
Caution: NOTE: CF™594 dCTP is not compatible with PCR labeling of DNA probes.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty