Would you like to log in again?
CF®DI Aminooxy, Biotium
Supplier: Biotium
92179 92177 92178
10001-668EA 268.24 USD InStock
10001-668 10001-666 10001-670
CF®DI Aminooxy, Biotium
Protein Linking and Labeling Systems
CF®DI aminooxy dyes react with aldehyde/ketone groups on proteins or other biomolecules to form a stable oxime bond. The CF®DI dyes (CF®488DI, CF®555DI and CF®647DI) are designed to match each other for molecular weight and charge. The molecular weights of the same protein conjugated to different CF®DI dye colors are close to each other and the pl of the labeled proteins remains the same.
  • React with carbonyl moieties of aldehyde or ketone groups on proteins and other biomolecules.
  • Size- and charge-matched fluorescent aminooxy dyes.
  • Available in 3 colors for multiplex analysis.
Biotium also offer original CF® Dye Aminooxy with a wide selection of colors. CF® dyes are next-generation fluorescent dyes with advantages in brightness, photostability, and water solubility compared to other fluorescent dyes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty