Would you like to log in again?
CF®DI Hydrazide, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
92184 92165 92164 92185 92166
75784-102EA 230.43 USD InStock
75784-102 10001-664 10001-660 10001-772 10001-662
CF®DI Hydrazide, Biotium
Protein Linking and Labeling Systems
CF®DI dye hydrazides react with molecules containing aldehyde or ketone groups to form a hydrazone bond. The dyes are size- and charge-matched and do not significantly alter the molecular weight or pI of the labeled proteins. They are used for detection of protein carbonylation by fluorescence 2D-DIGE.
  • React with carbonyl moieties of aldehyde or ketone groups.
  • Size- and charge-matched, do not significantly alter the molecular weight or pI of the labeled protein.
  • For detection of protein carbonylation by fluorescence 2D-DIGE.
  • Available in several colors for detection of multiple samples on the same gel.
Also see our regular CF® Dye Hydrazides, available in a wide selection of colors.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty