Would you like to log in again?
Cellular Senescence Live Cell Analysis Assay, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT130-0010
10001-774EA 429 USD InStock
10001-774
Cellular Senescence Live Cell Analysis Assay, Enzo Life Sciences
Assays Cellular Assays
Simple assay to quantify senescence-associated β-galactosidase activity, in live cells, using flow cytometry or fluorescent microscopy.
For the study of cellular senescence in live cells.
Assay kit that uses a fluorogenic substrate to measure SA-β-gal activity in live cells.
Ordering information: Contains Cell pretreatment solution (1000x) and SA-?-gal Substrate (200x).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty