Would you like to log in again?
Mini 500 Power Supply, G-Biosciences
Supplier: G-Biosciences
BT404
10001-824EA 1612.8 USD InStock
10001-824
Mini 500 Power Supply, G-Biosciences
Power Supplies
Mini 500 is made of polycarbonate housing and aluminum base plates which is compact sized and lightweight.
  • Safety Device: No load detection; shrouded plugs and sockets
  • Continuous or timed operation (1 to 999 minutes) with alarm function
  • Easily performed using the simple and user-friendly interface
The Mini 500 power supply is capable of providing constant voltage(<500 V) or constant current (<400 mA) in 1 V or 1 mA steps, and it is suited to run both vertical polyacrylamide or horizontal agarose gel electrophoresis experiments. The Mini 500 power supply features two electrode pairs, allowing for two gels to be run simultaneously, saving both time and valuable bench space.
Certifications: CE, ETL and FCC Marked.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty