Would you like to log in again?
DuoSource™ 150 Power Supplies, G-Biosciences
Supplier: G-Biosciences
BT401
10001-842EA 881.59 USD InStock
10001-842
DuoSource™ 150 Power Supplies, G-Biosciences
Power Supplies
The DuoSource™ 150 runs gels quickly (20 to 30 minutes at 150 V).
It is designed and affordable for classroom applications.
Certifications: CE marked
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty