Would you like to log in again?
HyFlex® 11-724 Medium Duty, 13-Gauge Gloves, Palm Dipped, Ansell
Supplier: Ansell
HyFlex®
11724060 11724080 11724090 11724110 11724070
10002-048CS 1994.06 USD InStock
10002-048 10001-966 10002-052 10002-050 10002-054
HyFlex® 11-724 Medium Duty, 13-Gauge Gloves, Palm Dipped, Ansell
Gloves
Excellent flexibility with optimal cut protection, the HyFlex® 11-724 glove features a 13G lightweight, medium-duty liner with INTERCEPT TECHNOLOGY® yarns that provide EN Cut Level 3 (ANSI Level 2) protection.
  • Silicone-free
  • Increased worker confidence when handling small, sharp parts
  • Exceptional abrasion resistance
  • Minimal shrinkage after laundering
The thin, ergonomic liner features an optimized Lycra® content that is soft and cool on the skin for all-day comfort.
Precision, dexterity, and flexibility when handling small, sharp parts increases worker confidence while on the job. Durable and launderable with minimal shrinkage and excellent recovery after laundering, extending glove life.
INTERCEPT TECHNOLOGY® yarn has been tested to perform at or exceed industry standards. This Ansell proprietary technology offers cut protection and increased comfort and dexterity. The palm coating design improves worker grip, extends glove life in abrasive conditions and allows the back of hand to breathe.
For use in aerospace, automotive, machinery and equipment and metal fabrication industries for assembly of small sharp parts, handling small sharp metal parts, light-duty stamping and maintenance.
Certifications: ANSI Cut Level 2. ANSI Abrasion Level 6.
Packaging: Packaged 12 pairs in a bag; 12 bags in a carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty