Would you like to log in again?
HyFlex® 11-541 Ultralight Weight, 18-Gauge Gloves, Palm Dipped, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyFlex®
11541080 11541060 11541100 11541070 11541100 11541110 11541060 11541080 11541070 11541110 11541090 11541090
10002-004CS 3852.51 USD InStock
10002-004 CA10002-078 10002-080 CA10002-004 10002-074 CA10002-082 CA10002-080 10002-082 CA10002-076 10002-078 10002-076 CA10002-074
HyFlex® 11-541 Ultralight Weight, 18-Gauge Gloves, Palm Dipped, Ansell
Gloves
Offering exceptional cut protection in a form-fitting ultralight weight glove with enhanced grip performance, the HyFlex® 11-541 epitomizes the convergence of protection and performance.
  • Enhanced abrasion resistance
  • Excellent grip
  • Superior tactile sensitivity
  • Unparalleled dexterity
It features an 18G ultra-lightweight liner with INTERCEPT TECHNOLOGY® yarns that provides EN Cut Level 4 (ANSI Cut Level 4) protection.
This glove's ultralight weight enables unparalleled dexterity and refined mobility for bare hand-like tactility. ZONZ™ comfort fit technology uses selected yarns and varying stitch designs to tailor overall glove fit to enhance hand movement for higher dexterity and reduced fatigue. Its silicone-free design is paint- and finish-process-friendly and dirt-masking palm allows longer wear times.
INTERCEPT TECHNOLOGY® yarn has been designed to perform at or exceed industry standards. This Ansell proprietary technology offers best-in-class cut protection and increased comfort with dexterity. Palm coated protection provides grip while not limiting comfort, and back-of-hand breathability. ANSELL GRIP™ technology is a coating treatment that minimizes the force required to grip oily or wet tools or materials, reducing hand and arm fatigue and improving control, safety and productivity. The HyFlex® 11-541 glove also features FORTIX™ technology which applies a thin, resilient and breathable nitrile foam coating, which extends gloves working life and improves comfort in abrasive applications.
For use in the aerospace, automotive, machinery and equipment, and metal fabrication industries for assembly of small sharp parts, handling small sharp metal parts, light duty stamping and maintenance.
Certifications: ANSI Cut Level 4. ANSI Abrasion Level 4. CE 0493 marked. EN 388:2003 - 4412. EN 388:2016 - 4X21D. EN 407 - X1XXXX. EN 1149. REACH Compliant.
Packaging: Packaged 12 pairs in a bag; 12 bags in a carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty