Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MME Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SN5c]

Supplier: Tonbo Biosciences
20-0108-T025 20-0108-T500 20-0108-T100
10002-104EA 240 USD InStock
10002-104 10002-102 10002-106
Anti-MME Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SN5c]
Antibodies
The SN5c antibody is specific for human CD10, a 100 kDa type II transmembrane protein also known as Common Acute Lymphoblastic Leukemia Antigen (CALLA). In normal cells, it is expressed on early B and T lymphoid precursors, neutrophils and on various epithelia. CD10 is also expressed on several lymphoma cells including Burkitts, acute lymphoblastic, and follicular germinal center lymphomas. CD10 functions as an endopeptidase and is involved in B cell development, as well as a mediator of neutrophil inflammatory responses.

Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: SN5c
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty