Would you like to log in again?
VerityPfu™ DNA Polymerase, Tonbo Biosciences
Supplier: Tonbo Biosciences
VerityPfu™
31-5020-1000U 31-5020-0200U
10002-214EA 1100 USD InStock
10002-214 10002-212
VerityPfu™ DNA Polymerase, Tonbo Biosciences
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VerityPfu DNA Polymerase is a proprietary next generation, high fidelity, proofreading enzyme that does not require hot start conditions
VerityPfu enzyme possesses exceptional proofreading activity that results in 50X higher fidelity than standard Taq DNA polymerase. For added convenience and value, the advanced Reaction Buffer (5X) includes dNTPs and MgCl2. PCR products generated with this enzyme are blunt-end finished.
Ultra-fast VerityPfu DNA Polymerase accurately and reliably generate amplicons up to 10 kb from complex genomic, viral, and plasmid templates.
Novel point mutations resulted in a Pfu variant with enhanced DNA binding that allows for significantly improved processing, yields and shortened cycle times.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty