Would you like to log in again?
FastPROBE™ qPCR Hi-ROX Master Mix, Tonbo Biosciences
Supplier: Tonbo Biosciences
FastPROBE™
31-5112-0100R 31-5112-0500R
10002-242EA 307 USD InStock
10002-242 10002-240
FastPROBE™ qPCR Hi-ROX Master Mix, Tonbo Biosciences
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
FastPROBE qPCR Hi-ROX Master Mix is ready-to-use cocktail for the amplification and quantitation of DNA on real-time instruments that support normalization with a standard ROX passive reference dye at a final concentration of 500 nM.
An integral component, the Classic++ Hot Start Taq DNA Polymerase is compatible with all fluorogenic probes including hydrolysis and displacement probes such as TaqMan®, Scorpions® and Molecular Beacons probes. Novel hot start technology prevents primer dimer formation and non-specific amplification resulting in higher yields and specificity. Advanced buffer chemistry means little to no protocol optimization, regardless of template.
FastPROBE qPCR master mixes are suitable for both standard and fast-cycling real-time PCR applications, providing improved signal to noise ratios, rapid extension rates for early Ct values, and enhanced sensitivity. Use for low copy number detection, microarray validation, SNP genotyping and multiplexing - Please refer to our Instrument Compatibility Guide to ensure you have the correct ROX concentration for your system.
Caution: Storage temperature: At -20°C upon Arrival and Limit Exposure to Light
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty