Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HB7]

Supplier: Tonbo Biosciences
50-0388-T025 50-0388-T100 50-0388-T500
10002-300EA 1110 USD InStock
10002-300 10002-298 10002-296
Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HB7]
Antibodies
The HB7 antibody is specific for human CD38, a 45 kDa type II transmembrane glycoprotein expressed on thymocytes, plasma cells, and monocytes as well as other non-hematopoietic cells. T and B lymphocyte expression is discontinuous and varies based on differentiation state � present on immature cells, low on intermediate stages, and high on activated mature cells. It is found on most CD34+ cells, but not on pluripotent stem cells. CD38 is an ectoenzyme that functions to catalyze the synthesis and hydrolysis of cyclic ADP-ribose and is involved in cell signaling, regulating cell adhesion, and activation. Additionally, CD38 has been shown to be a prognostic marker for some leukemias and other diseases.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HB7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty