Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: MY31]

Supplier: Tonbo Biosciences
60-0564-T100 60-0564-T025
10002-322EA 192 USD InStock
10002-322 10002-320
Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: MY31]
Antibodies
The MY31 antibody reacts with human CD56, also known as the neural cell adhesion molecule (NCAM), a glycoprotein which is a member of the immunoglobulin superfamily. The 140 kDa isoform of CD56 is expressed on human NK cells and NK-T cells, with increased expression levels on activated NK lymphocytes. The CD56 antigen is also expressed by neurons and is reported to play a role in nervous system development and neural cell-to-cell adhesion.

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: MY31
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty