Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Basic T Lymphocyte Backbone Panel [clone: 17A2, RM4-5, 53-6.7, PC61.5, IM7, MEL-14]Basic T Lymphocyte Backbone Panel

Supplier: Tonbo Biosciences
91-0002-U100 91-0002-U025
10002-346EA 705 USD InStock
10002-346 10002-344
Basic T Lymphocyte Backbone Panel [clone: 17A2, RM4-5, 53-6.7, PC61.5, IM7, MEL-14]Basic T Lymphocyte Backbone Panel
Antibodies
The Mouse Basic T Lymphocyte backbone panel is a six-color combination of anti-mouse antibodies. This panel can serve as a backbone for identifying various T lymphocyte subsets, including naive and memory T cell populations. This panel's configuration leaves room for GFP or additional drop-in markers, if desired. Violet laser (405 nm) required for detection of CD3e violetFluorTM 450

Contains:
1 x violetFluor™ 450 Anti-Mouse CD3 (17A2)
1 x APC-Cyanine7 Anti-Mouse CD4 (RM4-5)
1 x redFluor™ 710 Anti-Mouse CD8a (53-6.7)
1 x PE-Cyanine7 Anti-Mouse CD25 (PC61.5)
1 x PerCP-Cyanine5.5 Anti-Human/Mouse CD44 (IM7)
1 x APC Anti-Mouse CD62L (L-Selectin) (MEL-14)

Type: Primary
Antigen: CD3, CD4, CD8a; CD25, CD44, CD62L
Clonality: Monoclonal
Clone: 17A2, RM4-5, 53-6.7, PC61.5, IM7, MEL-14
Conjugation: violetFluor® 450, APC (Allophycocyanin)-Cy7, redFluor® 710, PE-Cy7, PerCP-Cy5.5, APC (Allophycocyanin)
Epitope:
Host:
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty