Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Myeloid Precursor Backbone Panel [clone: 17A2, M1/70, N418, 1D3, RA3-6B2, RB6-8C5, TER-119]Myeloid Precursor Backbone Panel

Supplier: Tonbo Biosciences
91-0003-U100 91-0003-U025
10002-348EA 411 USD InStock
10002-348 10002-350
Myeloid Precursor Backbone Panel [clone: 17A2, M1/70, N418, 1D3, RA3-6B2, RB6-8C5, TER-119]Myeloid Precursor Backbone Panel
Antibodies
The Mouse Myeloid Precursor backbone panel is a five-color combination of seven anti-mouse antibodies. This panel can serve as a backbone to identify various myeloid precursor populations, and leaves room for GFP or additional drop-in markers, if desired. The CD3e, CD19 and CD45R/B220 positive cells are gated together to exclude mature T and B cells. Violet laser (405 nm) required for detection of CD11b violetFluorTM 450. Hosts: Rat, Armenian Hamster

Contains:
1 x redFluor™ 710 Anti-Mouse CD3 (17A2)
1 x violetFluor™ 450 Anti-Human/Mouse CD11b (M1/70)
1 x PerCP-Cyanine5.5 Anti-Mouse CD11c (N418)
1 x redFluor™ 710 Anti-Mouse CD19 (1D3)
1 x redFluor™ 710 Anti-Human/Mouse CD45R (B220) (RA3-6B2)
1 x PE-Cyanine7 Anti-Mouse Ly-6G (Gr-1) (RB6-8C5)
1 x FITC Anti-Mouse TER-119 (TER-119)

Type: Primary
Antigen: CD3, CD11b, CD11c, CD19, CD45R, Ly-6G, TER-119
Clonality: Monoclonal
Clone: 17A2, M1/70, N418, 1D3, RA3-6B2, RB6-8C5, TER-119
Conjugation: redFluor® 710, violetFluor® 450, PerCP-Cy5.5, PE-Cy7, FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Multiple
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty