Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate), violetFluor® 450, PE, PerCP-Cy5.5) [clone: UCHT1, RPA-T4, RPA-T8, 2D1]

Supplier: Tonbo Biosciences
91-0101-T025 91-0101-T100
10002-360EA 260 USD InStock
10002-360 10002-362
Anti-CD3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate), violetFluor® 450, PE, PerCP-Cy5.5) [clone: UCHT1, RPA-T4, RPA-T8, 2D1]
Antibodies
The Human Basic T Lymphocyte backbone panel is a four-color combination of anti-human antibodies. This panel can serve as a backbone for identifying the major T cell subsets, and leaves room for additional drop-in markers, if desired. Violet laser (405 nm) required for detection of CD4 violetFluorTM 450.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1, RPA-T4, RPA-T8, 2D1
Conjugation: FITC (Fluorescein Isothiocyanate), violetFluor® 450, PE, PerCP-Cy5.5
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty