Would you like to log in again?
Peridox® Concentrate, Contec®
Supplier: Contec
CR85342NK
10002-762CS 724.4 USD InStock
10002-762
Peridox® Concentrate, Contec®
Cleaning Agents Disinfectants
EPA-registered Peridox® Concentrate is a disinfectant and cleaner that was designed to eliminate and prevent the spread of spores, as well as more than 40 other pathogens. This broad spectrum disinfectant is fast acting and is effective against bacterial spores in 3 minutes, bacteria and viruses in 2 minutes, fungus in 1 minute, and TB in 5 minutes. Peridox® Concentrate has more than 40 EPA registered claims and does not contain bleach, alcohol, phenols, or quaternary ammonium compounds.
  • Patented, hyperactive chemistry provides fast results, 3-minute sporicide claim at 99.9999% efficacy
  • EPA-registered disinfectant and cleaner is ideal for use in life science cleanrooms
  • Concentrated formula provides cleanroom facilities with a cost-effective approach to use a high-level sporicidal disinfectant facility-wide
  • Compatible with most cleanroom materials of construction, and can be used on workstations, stainless steel equipment, floors, walls, and ceilings
  • Does not contain bleach, alcohol, phenols or quaternary ammonium compounds
Peridox® Concentrate is fast acting and will not damage stainless steel equipment. Compatible with all Contec mopping systems and hardware. It requires no rinsing and leaves no film on surfaces, making it ideal for use on critical surfaces. With its shorter contact times and powerful wetting agents, Peridox® Concentrate stays wet on the surface longer and works faster.
Peridox® Concentrate does not contain bleach and will not damage clothing, carpet, upholstery, LCD screens, and other surfaces when used as directed.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty