Would you like to log in again?
Mouse Recombinant Caspase 1 (from E. coli)
Supplier: BioVision
AB52079-100
10005-120EA 425.26 USD InStock
10005-120
Mouse Recombinant Caspase 1 (from E. coli)
Enzymes
A cysteine protease involved in apoptosis 
Caspase-1 (also know as ICE) is a prototypical member of the caspase-family of cysteine proteases
The recombinant active mouse caspase-1 was expressed in E. coli. The active caspase-1 preferentially cleaves caspase-1 substrates (e.g., YVAD-AFC or YVAD-pNA) and is routinely tested at BioVision for its ability to enzymatically cleave these two substrates Ac-YVAD-pNA (Cat. #1104-200) or Ac-YVAD-AFC (1103-200).
Caspase-1 exists in cells as an inactive 45 kDa proenzyme. The pro-enzyme is matured by proteolysis to yield large (20 kD) and small (10 kD) subunits. The active caspase-1 is a heterotetramer consisting of two large and two small subunits. To date the regulatory mechanism of caspase-1 activation and the role of caspase-1 in apoptosis are poorly understood. In THP-1 cells, a large proportion of the caspase-1 is present in the inactive proenzyme form.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty