Would you like to log in again?
Human Recombinant IL15 (from E. coli)
Supplier: BioVision
AB285529-10UG AB285529-1MG AB285529-50UG
10005-638EA 2145.06 USD InStock
10005-638 10005-636 10006-242
Human Recombinant IL15 (from E. coli)
Proteins and Peptides
An inducer and mediator of proinflammatory responses.
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of CTLL-2 cells is <0.5 ng/ml, corresponding to a specific activity of >2 x 10⁶ units/mg.
IL-15 is a potent lymphoid cell growth factor that exerts its biological activity primarily on T cells. Human IL-15 is a 12.8 kDa protein containing 114 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty